2-(3-Phenyl[1,2,4]oxadiazol-5-yl)ethylamine hydrochloride

Supplier: Matrix Scientific

026433-500MG
101822-260EA 306.15 USD
101822-260
2-(3-Phenyl[1,2,4]oxadiazol-5-yl)ethylamine hydrochloride
2-(3-Phenyl[1,2,4]oxadiazol-5-yl)ethylamine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₂ClN₃O
MDL Number: MFCD09997587
CAS Number: 1185304-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR