3-(2-Hydroxymethylbenzoimidazol-1-yl)propionicacid hydrochloride

Supplier: Matrix Scientific

026430-500MG
101822-254EA 306.15 USD
101822-254
3-(2-Hydroxymethylbenzoimidazol-1-yl)propionicacid hydrochloride
3-(2-Hydroxymethylbenzoimidazol-1-yl)propionicacid hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O₃
MDL Number: MFCD09997585
CAS Number: 1052533-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR