Piperazine-1-carboxylic acid amide hydrochloride

Supplier: Matrix Scientific

Synonyms: Piperazine-1-carboxamide hydrochloride

026413-500MG
101822-226EA 307.69 USD
101822-226
Piperazine-1-carboxylic acid amide hydrochloride
Piperazine-1-carboxylic acid amide hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClN₃O
MW: 165.62 g/mol
MDL Number: MFCD06408948
CAS Number: 474711-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR