2-Furan-2-yl-1H-benzoimidazole-5-carboxylic acid hydrochloride

Supplier: Matrix Scientific

026387-500MG
101822-190EA 306.15 USD
101822-190
2-Furan-2-yl-1H-benzoimidazole-5-carboxylic acid hydrochloride
2-Furan-2-yl-1H-benzoimidazole-5-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₁₂H₉ClN₂O₃
MDL Number: MFCD03779940
CAS Number: 1158563-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR