3-Phenylpyrrolidine hydrochloride

Supplier: Matrix Scientific

026365-500MG
101822-154EA 95.38 USD
101822-154
3-Phenylpyrrolidine hydrochloride
3-Phenylpyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄ClN
MDL Number: MFCD02179094
CAS Number: 857281-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR