2-Piperazin-1-ylquinoline-4-carboxylic acid hydrochloride

Supplier: Matrix Scientific

026327-500MG
101822-092EA 236.92 USD
101822-092
2-Piperazin-1-ylquinoline-4-carboxylic acid hydrochloride
2-Piperazin-1-ylquinoline-4-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₆ClN₃O₂
MDL Number: MFCD09997543
CAS Number: 1185299-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR