3-Imidazol-1-ylpropionic acid hydrochloride

Supplier: Matrix Scientific

026325-500MG
101822-090EA 252.31 USD
101822-090
3-Imidazol-1-ylpropionic acid hydrochloride
3-Imidazol-1-ylpropionic acid hydrochloride

Hazard: Irritant

Formula: C₆H₉ClN₂O₂
MDL Number: MFCD05884968
CAS Number: 18999-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR