2-Cyclopropylaniline

Supplier: Matrix Scientific

Synonyms: 2-Cyclopropyl-phenylamine

026312-500MG
101822-064EA 500 USD
101822-064
2-Cyclopropylaniline
2-Cyclopropylaniline

Hazard: Irritant

Formula: C₉H₁₁N
MW: 133.19 g/mol
MDL Number: MFCD00604193
CAS Number: 3158-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR