1-Ethyl-2-methyl-1H-benzoimidazol-5-ylamine dihydrochloride

Supplier: Matrix Scientific

026310-500MG
101822-060EA 306.15 USD
101822-060
1-Ethyl-2-methyl-1H-benzoimidazol-5-ylamine dihydrochloride
1-Ethyl-2-methyl-1H-benzoimidazol-5-ylamine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅Cl₂N₃
MDL Number: MFCD00035184
CAS Number: 73688-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR