2-tert-Butyl-4-[3-(dimethylvinylsilanyl)propoxy]-6-(5-methoxybenzotriazol-2-yl)phenol

Supplier: Matrix Scientific

026292-500MG
101822-002EA 2520 USD
101822-002
2-tert-Butyl-4-[3-(dimethylvinylsilanyl)propoxy]-6-(5-methoxybenzotriazol-2-yl)phenol
2-tert-Butyl-4-[3-(dimethylvinylsilanyl)propoxy]-6-(5-methoxybenzotriazol-2-yl)phenol

Hazard: Irritant

Formula: C₂₄H₃₃N₃O₃Si
MDL Number: MFCD09763676
CAS Number: 122430-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR