tert-Butyl-4-(5-chloro-1H-pyrrolo[2,3-b]pyridin-4-yl)piperazine-1-carboxylate

Supplier: Matrix Scientific

026289-100MG
101821-996EA 196.92 USD
101821-996
tert-Butyl-4-(5-chloro-1H-pyrrolo[2,3-b]pyridin-4-yl)piperazine-1-carboxylate
tert-Butyl-4-(5-chloro-1H-pyrrolo[2,3-b]pyridin-4-yl)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₁ClN₄O₂
MW: 336.82 g/mol
MDL Number: MFCD09965901
CAS Number: 1020056-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR