5-Chloro-4-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)-1-triisopropylsilanyl-1H-pyrrolo[2,3-b]pyridine

Supplier: Matrix Scientific

026288-100MG
101821-994EA 133.85 USD
101821-994
5-Chloro-4-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)-1-triisopropylsilanyl-1H-pyrrolo[2,3-b]pyridine
5-Chloro-4-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)-1-triisopropylsilanyl-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₂₂H₃₆BClN₂O₂Si
MDL Number: MFCD09965900
CAS Number: 1072152-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR