5-Chloro-1H-pyrrolo[2,3-b]pyridin-4-ol

Supplier: Matrix Scientific

026282-100MG
101821-982EA 364.62 USD
101821-982
5-Chloro-1H-pyrrolo[2,3-b]pyridin-4-ol
5-Chloro-1H-pyrrolo[2,3-b]pyridin-4-ol

Hazard: Irritant

Formula: C₇H₅ClN₂O
MDL Number: MFCD09965893
CAS Number: 1015610-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR