5-Chloro-4-methoxy-1H-pyrrolo[2,3-b]pyridine
Supplier: Matrix Scientific
026278-100MG
101821-974EA
364.62
USD
101821-974
5-Chloro-4-methoxy-1H-pyrrolo[2,3-b]pyridine
5-Chloro-4-methoxy-1H-pyrrolo[2,3-b]pyridine
Hazard: Irritant
Formula:
C₈H₇ClN₂O |
MDL Number:
MFCD09965888 CAS Number: 1020056-69-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...