4-Chloro-5-iodo-1H-pyrrolo[2,3-b]pyridine

Supplier: Matrix Scientific

026276-100MG
101821-970EA 364.62 USD
101821-970
4-Chloro-5-iodo-1H-pyrrolo[2,3-b]pyridine
4-Chloro-5-iodo-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₇H₄ClIN₂
MDL Number: MFCD09965886
CAS Number: 1015610-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR