5-Fluoro-4-iodo-1H-pyrrolo[2,3-b]pyridine

Supplier: Matrix Scientific

026275-100MG
101821-968EA 229.23 USD
101821-968
5-Fluoro-4-iodo-1H-pyrrolo[2,3-b]pyridine
5-Fluoro-4-iodo-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₇H₄FIN₂
MDL Number: MFCD09965885
CAS Number: 1015610-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR