(4-Chloro-1H-pyrrolo[2,3-b]pyridin-5-yl)methanol

026271-100MG
101821-960EA 256.61 USD
101821-960
(4-Chloro-1H-pyrrolo[2,3-b]pyridin-5-yl)methanol
(4-Chloro-1H-pyrrolo[2,3-b]pyridin-5-yl)methanol

Hazard: Irritant

Formula: C₈H₇ClN₂O
MDL Number: MFCD09965880
CAS Number: 1015610-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR