4-Chloro-1H-pyrrolo[2,3-b]pyridine-5-carbaldehyde

Supplier: Matrix Scientific

026270-100MG
101821-958EA 43.08 USD
101821-958
4-Chloro-1H-pyrrolo[2,3-b]pyridine-5-carbaldehyde
4-Chloro-1H-pyrrolo[2,3-b]pyridine-5-carbaldehyde

Hazard: Irritant

Formula: C₈H₅ClN₂O
MDL Number: MFCD09859124
CAS Number: 958230-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR