Methyl-5-chloro-1H-pyrrolo[2,3-b]pyridine-4-carboxylate

Supplier: Matrix Scientific

026269-100MG
101821-956EA 243.08 USD
101821-956
Methyl-5-chloro-1H-pyrrolo[2,3-b]pyridine-4-carboxylate
Methyl-5-chloro-1H-pyrrolo[2,3-b]pyridine-4-carboxylate

Hazard: Irritant

Formula: C₉H₇ClN₂O₂
MDL Number: MFCD09965879
CAS Number: 1015609-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR