tert-Butyl-4-(1H-pyrrolo[2,3-b]pyridin-4-yl)piperazine-1-carboxylate

026266-100MG
101821-950EA 328.64 USD
101821-950
tert-Butyl-4-(1H-pyrrolo[2,3-b]pyridin-4-yl)piperazine-1-carboxylate
tert-Butyl-4-(1H-pyrrolo[2,3-b]pyridin-4-yl)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₂N₄O₂
MW: 302.38 g/mol
MDL Number: MFCD09859122
CAS Number: 577768-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR