6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[3,2-b]pyridine

Supplier: Matrix Scientific

026264-100MG
101821-946EA 244.62 USD
101821-946
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[3,2-b]pyridine
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[3,2-b]pyridine

Hazard: Irritant

Formula: C₁₃H₁₇BN₂O₂
MDL Number: MFCD09859120
CAS Number: 1045855-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR