(1H-Pyrrolo[3,2-b]pyridin-6-yl)methanol

Supplier: Matrix Scientific

026259-100MG
101821-936EA 223.08 USD
101821-936
(1H-Pyrrolo[3,2-b]pyridin-6-yl)methanol
(1H-Pyrrolo[3,2-b]pyridin-6-yl)methanol

Hazard: Irritant

Formula: C₈H₈N₂O
MDL Number: MFCD09859116
CAS Number: 1015609-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR