4-Chloro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine

Supplier: Matrix Scientific

026254-100MG
101821-926EA 364.62 USD
101821-926
4-Chloro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine
4-Chloro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₁₆H₂₅ClN₂Si
MDL Number: MFCD09763673
CAS Number: 651744-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR