1H-Pyrrolo[3,2-b]pyridine-6-carboxylic acid

026251-100MG
101821-920EA 394.88 USD
101821-920
1H-Pyrrolo[3,2-b]pyridine-6-carboxylic acid
1H-Pyrrolo[3,2-b]pyridine-6-carboxylic acid

Hazard: Irritant

Formula: C₈H₆N₂O₂
MDL Number: MFCD09763670
CAS Number: 112766-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR