3-Iodo-1H-pyrrolo[2,3-b]pyridine-4-carbonitrile

Supplier: Matrix Scientific

026248-100MG
101821-914EA 115.38 USD
101821-914
3-Iodo-1H-pyrrolo[2,3-b]pyridine-4-carbonitrile
3-Iodo-1H-pyrrolo[2,3-b]pyridine-4-carbonitrile

Hazard: Irritant

Formula: C₈H₄IN₃
MDL Number: MFCD09763667
CAS Number: 956485-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR