1H-Pyrrolo[2,3-b]pyridine-5-carboxylic acid methoxymethylamide

Supplier: Matrix Scientific

026238-100MG
101821-894EA 364.62 USD
101821-894
1H-Pyrrolo[2,3-b]pyridine-5-carboxylic acid methoxymethylamide
1H-Pyrrolo[2,3-b]pyridine-5-carboxylic acid methoxymethylamide

Hazard: Irritant

Formula: C₁₀H₁₁N₃O₂
MDL Number: MFCD09763663
CAS Number: 944936-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR