4-Methoxy-1-triisopropylsilanyl-1H-pyrrolo[2,3-b]pyridine

Supplier: Matrix Scientific

026233-100MG
101821-880EA 364.62 USD
101821-880
4-Methoxy-1-triisopropylsilanyl-1H-pyrrolo[2,3-b]pyridine
4-Methoxy-1-triisopropylsilanyl-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₁₇H₂₈N₂OSi
MDL Number: MFCD08741548
CAS Number: 944936-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR