4-Chloro-1H-pyrrolo[2,3-b]pyridine-5-carbonitrile
Supplier: Matrix Scientific
026231-100MG
101821-876EA
244.62
USD
101821-876
4-Chloro-1H-pyrrolo[2,3-b]pyridine-5-carbonitrile
4-Chloro-1H-pyrrolo[2,3-b]pyridine-5-carbonitrile
Hazard: Irritant
Formula:
C₈H₄ClN₃ |
MDL Number:
MFCD09965877 CAS Number: 920966-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...