5-Methoxypyrrolo[2,3-b]pyridine-1-carboxylic acid tert-butyl ester

Supplier: Matrix Scientific

026230-100MG
101821-874EA 364.62 USD
101821-874
5-Methoxypyrrolo[2,3-b]pyridine-1-carboxylic acid tert-butyl ester
5-Methoxypyrrolo[2,3-b]pyridine-1-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₃
MW: 248.28 g/mol
MDL Number: MFCD08741547
CAS Number: 928653-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR