5-Bromo-3-iodopyrrolo[2,3-b]pyridine-1-carboxylic acid tert-butyl ester

Supplier: Matrix Scientific

026218-100MG
101821-850EA 364.62 USD
101821-850
5-Bromo-3-iodopyrrolo[2,3-b]pyridine-1-carboxylic acid tert-butyl ester
5-Bromo-3-iodopyrrolo[2,3-b]pyridine-1-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₂H₁₂BrIN₂O₂
MW: 423.06 g/mol
MDL Number: MFCD08741535
CAS Number: 928653-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR