3-Iodo-1H-pyrrolo[2,3-b]pyridine-5-carbonitrile

Supplier: Matrix Scientific

026202-100MG
101821-816EA 244.62 USD
101821-816
3-Iodo-1H-pyrrolo[2,3-b]pyridine-5-carbonitrile
3-Iodo-1H-pyrrolo[2,3-b]pyridine-5-carbonitrile

Hazard: Irritant

Formula: C₈H₄IN₃
MDL Number: MFCD08457791
CAS Number: 757978-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR