5-Bromo-2-iodo-3-methoxypyridine

Supplier: Matrix Scientific

026196-250MG
101821-806EA 383.08 USD
101821-806
5-Bromo-2-iodo-3-methoxypyridine
5-Bromo-2-iodo-3-methoxypyridine

Hazard: Irritant

Formula: C₆H₅BrINO
MDL Number: MFCD09859111
CAS Number: 944805-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR