tert-Butyl-5-iodo-4-methoxypyridin-2-ylcarbamate

Supplier: Matrix Scientific

026195-250MG
101821-804EA 266.15 USD
101821-804
tert-Butyl-5-iodo-4-methoxypyridin-2-ylcarbamate
tert-Butyl-5-iodo-4-methoxypyridin-2-ylcarbamate

Hazard: Irritant

Formula: C₁₁H₁₅IN₂O₃
MW: 350.16 g/mol
MDL Number: MFCD09763662
CAS Number: 944935-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR