3-Amino-2,5-dibromopyridine

Supplier: Matrix Scientific

Synonyms: 2,5-Dibromopyridin-3-amine

026192-250MG
101821-798EA 464.62 USD
101821-798
3-Amino-2,5-dibromopyridine
3-Amino-2,5-dibromopyridine

Hazard: Irritant

Formula: C₅H₄Br₂N₂
MW: 251.9100 g/mol
MDL Number: MFCD09266224
CAS Number: 90902-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR