(2-Chloro-5-fluoropyridin-3-yl)methanol

Supplier: Matrix Scientific

026184-250MG
101821-776EA 429.23 USD
101821-776
(2-Chloro-5-fluoropyridin-3-yl)methanol
(2-Chloro-5-fluoropyridin-3-yl)methanol

Hazard: Irritant

Formula: C₆H₅ClFNO
MW: 161.56 g/mol
MDL Number: MFCD03095313
CAS Number: 870063-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR