3-(2-Amino-5-bromopyridin-3-yl)acrylic acidmethyl ester

Supplier: Matrix Scientific

026183-250MG
101821-774EA 300 USD
101821-774
3-(2-Amino-5-bromopyridin-3-yl)acrylic acidmethyl ester
3-(2-Amino-5-bromopyridin-3-yl)acrylic acidmethyl ester

Hazard: Irritant

Formula: C₉H₉BrN₂O₂
MDL Number: MFCD07781129
CAS Number: 912760-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR