N-(2-Methoxy-pyridin-4-yl)-2,2-dimethylpropionamide

Supplier: Matrix Scientific

026178-250MG
101821-762EA 303.08 USD
101821-762
N-(2-Methoxy-pyridin-4-yl)-2,2-dimethylpropionamide
N-(2-Methoxy-pyridin-4-yl)-2,2-dimethylpropionamide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MW: 208.26 g/mol
MDL Number: MFCD08688593
CAS Number: 898561-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR