N-(5-Dimethoxymethyl-pyridin-2-yl)-2,2-dimethylpropionamide

Supplier: Matrix Scientific

026174-250MG
101821-752EA 300 USD
101821-752
N-(5-Dimethoxymethyl-pyridin-2-yl)-2,2-dimethylpropionamide
N-(5-Dimethoxymethyl-pyridin-2-yl)-2,2-dimethylpropionamide

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₃
MW: 252.32 g/mol
MDL Number: MFCD08688590
CAS Number: 898561-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR