N-(5-Methoxypyridin-2-yl)-2,2-dimethylpropionamide

Supplier: Matrix Scientific

026173-250MG
101821-750EA 300 USD
101821-750
N-(5-Methoxypyridin-2-yl)-2,2-dimethylpropionamide
N-(5-Methoxypyridin-2-yl)-2,2-dimethylpropionamide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MW: 208.26 g/mol
MDL Number: MFCD08688589
CAS Number: 898561-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR