N-(5-Hydroxymethylpyridin-2-yl)-2,2-dimethylpropionamide

Supplier: Matrix Scientific

026170-250MG
101821-744EA 306.15 USD
101821-744
N-(5-Hydroxymethylpyridin-2-yl)-2,2-dimethylpropionamide
N-(5-Hydroxymethylpyridin-2-yl)-2,2-dimethylpropionamide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MW: 208.26 g/mol
MDL Number: MFCD08457784
CAS Number: 882016-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR