N-(5-Chloropyridin-3-yl)-2,2-dimethylpropionamide

Supplier: Matrix Scientific

026166-250MG
101821-736EA 298.46 USD
101821-736
N-(5-Chloropyridin-3-yl)-2,2-dimethylpropionamide
N-(5-Chloropyridin-3-yl)-2,2-dimethylpropionamide

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O
MW: 212.68 g/mol
MDL Number: MFCD08235141
CAS Number: 879326-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR