3-(5-Methoxypyridin-3-yl)prop-2-yn-1-ol

Supplier: Matrix Scientific

026161-250MG
101821-726EA 300 USD
101821-726
3-(5-Methoxypyridin-3-yl)prop-2-yn-1-ol
3-(5-Methoxypyridin-3-yl)prop-2-yn-1-ol

Hazard: Irritant

Formula: C₉H₉NO₂
MDL Number: MFCD08457778
CAS Number: 898561-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR