5-(2,2-Dimethylpropionylamino)nicotinic acid

Supplier: Matrix Scientific

026160-250MG
101821-724EA 183.08 USD
101821-724
5-(2,2-Dimethylpropionylamino)nicotinic acid
5-(2,2-Dimethylpropionylamino)nicotinic acid

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₃
MW: 222.25 g/mol
MDL Number: MFCD08235140
CAS Number: 879326-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR