5-Iodopyridin-3-ylamine

Supplier: Matrix Scientific

026158-250MG
101821-720EA 273.85 USD
101821-720
5-Iodopyridin-3-ylamine
5-Iodopyridin-3-ylamine

Hazard: Irritant

Formula: C₅H₅IN₂
MW: 220.01 g/mol
MDL Number: MFCD08235139
CAS Number: 25391-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR