N-(5-Iodopyridin-3-yl)-2,2-dimethylpropionamide

Supplier: Matrix Scientific

026157-250MG
101821-718EA 303.08 USD
101821-718
N-(5-Iodopyridin-3-yl)-2,2-dimethylpropionamide
N-(5-Iodopyridin-3-yl)-2,2-dimethylpropionamide

Hazard: Irritant

Formula: C₁₀H₁₃IN₂O
MW: 304.13 g/mol
MDL Number: MFCD08235138
CAS Number: 873302-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR