3-Dimethoxymethyl-5-trimethylsilanylethynyl pyridine

Supplier: Matrix Scientific

026156-250MG
101821-716EA 303.08 USD
101821-716
3-Dimethoxymethyl-5-trimethylsilanylethynyl pyridine
3-Dimethoxymethyl-5-trimethylsilanylethynyl pyridine

Hazard: Irritant

Formula: C₁₃H₁₉NO₂Si
MW: 249.39 g/mol
MDL Number: MFCD08457777
CAS Number: 879326-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR