(5-Iodo-pyridin-3-yl)methanol

Supplier: Matrix Scientific

026154-250MG
101821-712EA 275.38 USD
101821-712
(5-Iodo-pyridin-3-yl)methanol
(5-Iodo-pyridin-3-yl)methanol

Hazard: Irritant

Formula: C₆H₆INO
MDL Number: MFCD08457775
CAS Number: 72299-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR