N-(3-Iodo-4-methoxy-pyridin-2-yl)-2,2-dimethyl-propionamide

Supplier: Matrix Scientific

026150-250MG
101821-704EA 303.08 USD
101821-704
N-(3-Iodo-4-methoxy-pyridin-2-yl)-2,2-dimethyl-propionamide
N-(3-Iodo-4-methoxy-pyridin-2-yl)-2,2-dimethyl-propionamide

Hazard: Irritant

Formula: C₁₁H₁₅IN₂O₂
MDL Number: MFCD08235132
CAS Number: 898561-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR