N-(4-Chloro-3-iodopyridin-2-yl)-2,2-dimethylpropionamide

Supplier: Matrix Scientific

Synonyms: N-(4-Chloro-3-iodopyridin-2-yl)pivalamide

026149-250MG
101821-702EA 300 USD
101821-702
N-(4-Chloro-3-iodopyridin-2-yl)-2,2-dimethylpropionamide
N-(4-Chloro-3-iodopyridin-2-yl)-2,2-dimethylpropionamide

Hazard: Irritant

Formula: C₁₀H₁₂ClIN₂O
MW: 338.58 g/mol
MDL Number: MFCD08235131
CAS Number: 898561-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR