N-(3-[1,3]Dioxolan-2-ylpyridin-4-yl)-2,2-dimethylpropionamide

Supplier: Matrix Scientific

026144-500MG
101821-690EA 312.31 USD
101821-690
N-(3-[1,3]Dioxolan-2-ylpyridin-4-yl)-2,2-dimethylpropionamide
N-(3-[1,3]Dioxolan-2-ylpyridin-4-yl)-2,2-dimethylpropionamide

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₃
MW: 250.3 g/mol
MDL Number: MFCD06210003
CAS Number: 773087-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR